ENA413170430 Enamine

2,5-dichloroquinoline

  • MDL number MFCD00832074
  • Cas Number : 59412-12-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-152566

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-152566
Availability details for
Quantity
  • Availability
  • Add to Cart