ENA413170431 Enamine

4-amino-N,5-dimethyl-N-phenyl-1H-pyrazole-3-carboxamide

  • MDL number MFCD17298320
  • Cas Number : 1346268-79-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-152567

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-152567
Availability details for
Quantity
  • Availability
  • Add to Cart