ENA413170432 Enamine

4-amino-5-methyl-N-phenyl-1H-pyrazole-3-carboxamide

  • MDL number MFCD16676347
  • Cas Number : 178320-03-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-152568

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-152568
Availability details for
Quantity
  • Availability
  • Add to Cart