ENA413170435 Enamine

ethyl(2-phenylbutyl)amine

  • MDL number MFCD11204905
  • Cas Number : 402920-83-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-152572

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-152572
Availability details for
Quantity
  • Availability
  • Add to Cart