ENA413170437 Enamine

5-(1H-1,3-benzodiazol-2-yl)-2-fluoroaniline

  • MDL number MFCD12732357
  • Cas Number : 1183634-12-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-152574

Safety Information

msds https://www.enaminestore.com/catalog/EN300-152574
Availability details for
Quantity
  • Availability
  • Add to Cart