ENA413170441 Enamine

tert-butyl N-[3-(2-hydroxyethyl)pyridin-4-yl]carbamate

  • MDL number MFCD20491404
  • Cas Number : 219834-80-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-152582

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-152582
Availability details for
Quantity
  • Availability
  • Add to Cart