ENA413170455 Enamine

1-{[4-(trifluoromethyl)phenyl]methyl}pyrrolidin-3-ol

  • MDL number MFCD11048139
  • Cas Number : 1044769-56-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-152601

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-152601
Availability details for
Quantity
  • Availability
  • Add to Cart