ENA413170473 Enamine

N-(prop-2-yn-1-yl)cyclopropanamine

  • MDL number MFCD16480818
  • Cas Number : 49565-47-1
  • Molecular Weight : 95.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-151667

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-151667
Availability details for
Quantity
  • Availability
  • Add to Cart