ENA413170474 Enamine

N-[(3-bromophenyl)methyl]-N-methylaniline

  • MDL number MFCD14644634
  • Cas Number : 16547-15-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-152656

Safety Information

msds https://www.enaminestore.com/catalog/EN300-152656
Availability details for
Quantity
  • Availability
  • Add to Cart