ENA413170475 Enamine

N-[(3-bromophenyl)methyl]-N-methylcyclopropanamine

  • MDL number MFCD14648857
  • Cas Number : 1249199-38-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-152659

Safety Information

msds https://www.enaminestore.com/catalog/EN300-152659
Availability details for
Quantity
  • Availability
  • Add to Cart