ENA413170476 Enamine

tert-butyl N-{3-[methoxy(methyl)carbamoyl]pyridin-4-yl}carbamate

  • MDL number MFCD24474436
  • Cas Number : 244780-32-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-152661

Safety Information

msds https://www.enaminestore.com/catalog/EN300-152661
Availability details for
Quantity
  • Availability
  • Add to Cart