ENA413170477 Enamine

4-{[(tert-butoxy)carbonyl]amino}pyridine-3-carboxylic acid

  • MDL number MFCD03427724
  • Cas Number : 171178-34-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-152663

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-152663
Availability details for
Quantity
  • Availability
  • Add to Cart