ENA413170485 Enamine

N-(2-cyano-3-fluorophenyl)propanamide

  • MDL number MFCD12142487
  • Cas Number : 1155153-85-7
  • Molecular Weight : 192.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-152704

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-152704
Availability details for
Quantity
  • Availability
  • Add to Cart