ENA413170487 Enamine

5-tert-butyl 2-methyl 3-amino-4H,5H,6H-thieno[2,3-c]pyrrole-2,5-dicarboxylate

  • MDL number MFCD22690002
  • Cas Number : 1357948-71-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-152707

Safety Information

msds https://www.enaminestore.com/catalog/EN300-152707
Availability details for
Quantity
  • Availability
  • Add to Cart