ENA413170502 Enamine

2-chloro-6-methyl-3-nitropyridin-4-amine

  • MDL number MFCD08272055
  • Cas Number : 1026907-18-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-152735

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-152735
Availability details for
Quantity
  • Availability
  • Add to Cart