ENA413170517 Enamine

methyl 5-methyl-3H-imidazo[4,5-b]pyridine-7-carboxylate

  • MDL number MFCD20688525
  • Cas Number : 1378793-08-8
  • Molecular Weight : 191.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 242-243
Iupac Name methyl 5-methyl-3H-imidazo[4,5-b]pyridine-7-carboxylate
Inchi Code 1S/C9H9N3O2/c1-5-3-6(9(13)14-2)7-8(12-5)11-4-10-7/h3-4H,1-2H3,(H,10,11,12)
Inchi Key NDWZUOWRQVDIBY-UHFFFAOYSA-N

Description

Melting Point 242-243
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-152757

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P363,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-152757
pictograms GHS07
hstatements H302,H312,H315,H319,H332,H335
Availability details for
Quantity
  • Availability
  • Add to Cart