ENA413170521 Enamine

N-(1,3-thiazol-2-yl)piperidin-4-amine

  • MDL number MFCD11642828
  • Cas Number : 943330-52-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-152771

Safety Information

msds https://www.enaminestore.com/catalog/EN300-152771
Availability details for
Quantity
  • Availability
  • Add to Cart