ENA413170523 Enamine

2-(bromomethyl)-6-fluorobenzonitrile

  • MDL number MFCD18394042
  • Cas Number : 1261686-95-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-152774

Safety Information

msds https://www.enaminestore.com/catalog/EN300-152774
Availability details for
Quantity
  • Availability
  • Add to Cart