ENA413170526 Enamine

2,3-difluorobenzene-1-sulfonamide

  • MDL number MFCD12785794
  • Cas Number : 1133122-99-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-152779

Safety Information

msds https://www.enaminestore.com/catalog/EN300-152779
Availability details for
Quantity
  • Availability
  • Add to Cart