ENA413170528 Enamine

(2,3-dihydro-1H-isoindol-5-yl)methanamine dihydrochloride

  • MDL number MFCD27500782
  • Cas Number : 1797233-99-8
  • Molecular Weight : 221.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name isoindolin-5-ylmethanamine dihydrochloride
Inchi Code 1S/C9H12N2.2ClH/c10-4-7-1-2-8-5-11-6-9(8)3-7;;/h1-3,11H,4-6,10H2;2*1H
Inchi Key RGPMNXKUMWMQCJ-UHFFFAOYSA-N

Description

Purity 95
Salt Data Cl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-152798

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-152798
Availability details for
Quantity
  • Availability
  • Add to Cart