ENA413170530 Enamine

2-(4-chlorobenzenesulfinyl)acetic acid

  • MDL number MFCD14694623
  • Cas Number : 3996-47-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-152802

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-152802
Availability details for
Quantity
  • Availability
  • Add to Cart