ENA413170538 Enamine

(2S)-1-(benzyloxy)propan-2-amine

  • MDL number MFCD10033508
  • Cas Number : 83053-85-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-152846

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-152846
Availability details for
Quantity
  • Availability
  • Add to Cart