ENA413170539 Enamine

4-{5H,6H,7H,8H-[1,2,4]triazolo[4,3-a]pyridin-3-yl}piperidine

  • MDL number MFCD24036095
  • Cas Number : 1519824-36-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-152848

Safety Information

msds https://www.enaminestore.com/catalog/EN300-152848
Availability details for
Quantity
  • Availability
  • Add to Cart