ENA413170540 Enamine

4-[(1-{[1,2,4]triazolo[4,3-a]pyridin-3-yl}ethyl)carbamoyl]butanoic acid

  • MDL number MFCD27500786
  • Cas Number : 1184188-37-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-152849

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-152849
Availability details for
Quantity
  • Availability
  • Add to Cart