ENA413170543 Enamine

1,3-dimethyl-2-oxo-2,3-dihydro-1H-1,3-benzodiazole-5-sulfonamide

  • MDL number MFCD03197041
  • Cas Number : 24134-67-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-152863

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-152863
Availability details for
Quantity
  • Availability
  • Add to Cart