ENA413170545 Enamine

(2E)-N-(4-methoxyphenyl)imidazolidin-2-imine

  • MDL number MFCD03321714
  • Cas Number : 53976-95-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-152869

Safety Information

msds https://www.enaminestore.com/catalog/EN300-152869
Availability details for
Quantity
  • Availability
  • Add to Cart