ENA413170549 Enamine

4,6-dimethyl-1H-indole-2-carbaldehyde

  • MDL number MFCD24616650
  • Cas Number : 1258520-62-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-152884

Safety Information

msds https://www.enaminestore.com/catalog/EN300-152884
Availability details for
Quantity
  • Availability
  • Add to Cart