ENA413170560 Enamine

3-methyl-2,3,4,5-tetrahydro-1,4-benzothiazepine

  • MDL number MFCD20673000
  • Cas Number : 802875-04-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-152915

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-152915
Availability details for
Quantity
  • Availability
  • Add to Cart