ENA413170568 Enamine

1-(1,3-benzothiazol-6-yl)ethan-1-amine

  • MDL number MFCD12408400
  • Cas Number : 916201-65-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-152951

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-152951
Availability details for
Quantity
  • Availability
  • Add to Cart