ENA413170571 Enamine

1-(azepan-1-yl)propan-2-amine

  • MDL number MFCD09262751
  • Cas Number : 54151-46-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-152983

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-152983
Availability details for
Quantity
  • Availability
  • Add to Cart