ENA413170572 Enamine

2-{[(tert-butoxy)carbonyl]amino}-4-oxopentanoic acid

  • MDL number MFCD24135199
  • Cas Number : 160080-53-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-152984

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-152984
Availability details for
Quantity
  • Availability
  • Add to Cart