ENA413170575 Enamine

1-(2,3-dihydro-1H-indol-5-yl)ethan-1-amine

  • MDL number MFCD18646890
  • Cas Number : 765945-09-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-152987

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-152987
Availability details for
Quantity
  • Availability
  • Add to Cart