ENA413170578 Enamine

(2-amino-1-cyclopropylethyl)(benzyl)methylamine

  • MDL number MFCD12109265
  • Cas Number : 1156274-66-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-152993

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-152993
Availability details for
Quantity
  • Availability
  • Add to Cart