ENA413170580 Enamine

2-cyano-N-ethyl-N-phenylacetamide

  • MDL number MFCD09938464
  • Cas Number : 922178-99-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-152998

Safety Information

msds https://www.enaminestore.com/catalog/EN300-152998
Availability details for
Quantity
  • Availability
  • Add to Cart