ENA413170586 Enamine

5-(4-methylphenyl)-5-oxopentanenitrile

  • MDL number MFCD00464539
  • Cas Number : 88636-41-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-153004

Safety Information

msds https://www.enaminestore.com/catalog/EN300-153004
Availability details for
Quantity
  • Availability
  • Add to Cart