ENA413170588 Enamine

2-[benzyl(methyl)amino]-1-phenylpropan-1-one

  • MDL number MFCD09264308
  • Cas Number : 16735-30-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-153006

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-153006
Availability details for
Quantity
  • Availability
  • Add to Cart