ENA413170603 Enamine

(1R)-1-(oxan-4-yl)ethan-1-amine hydrochloride

  • MDL number MFCD19382414
  • Cas Number : 1269756-03-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-153036

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-153036
Availability details for
Quantity
  • Availability
  • Add to Cart