ENA413170604 Enamine

1-[5-(2-chlorophenyl)furan-2-carbonyl]-4-[2-(pyrrolidin-1-yl)ethyl]piperazine

  • MDL number MFCD09391489
  • Cas Number : 941047-27-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-153037

Safety Information

msds https://www.enaminestore.com/catalog/EN300-153037
Availability details for
Quantity
  • Availability
  • Add to Cart