ENA413170610 Enamine

tert-butyl N-(3-amino-2,2-difluoropropyl)carbamate

  • MDL number MFCD26516645
  • Cas Number : 1044675-84-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-153048

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-153048
Availability details for
Quantity
  • Availability
  • Add to Cart