ENA413170612 Enamine

4-(4-hydroxyphenoxy)-3-methylbenzonitrile

  • MDL number MFCD12872560
  • Cas Number : 1184486-24-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-153058

Safety Information

msds https://www.enaminestore.com/catalog/EN300-153058
Availability details for
Quantity
  • Availability
  • Add to Cart