ENA413170634 Enamine

1-[5-(3-fluorophenyl)-1,3,4-thiadiazol-2-yl]ethan-1-amine

  • MDL number MFCD21289437
  • Cas Number : 1505610-39-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-153136

Safety Information

msds https://www.enaminestore.com/catalog/EN300-153136
Availability details for
Quantity
  • Availability
  • Add to Cart