ENA413170636 Enamine

4-chloro-1-methyl-5-nitro-1H-pyrazole

  • MDL number MFCD00466344
  • Cas Number : 84547-95-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-153139

Safety Information

msds https://www.enaminestore.com/catalog/EN300-153139
Availability details for
Quantity
  • Availability
  • Add to Cart