ENA413170638 Enamine

5-(5-methylfuran-2-yl)-1,2-oxazole-4-carboxylic acid

  • MDL number MFCD24170067
  • Cas Number : 1469045-05-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-153145

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-153145
Availability details for
Quantity
  • Availability
  • Add to Cart