ENA413170642 Enamine

2-(2-ethoxyethoxy)ethyl carbamate

  • MDL number MFCD20708682
  • Cas Number : 1379303-63-5
  • Molecular Weight : 177.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(2-ethoxyethoxy)ethyl carbamate
Inchi Code 1S/C7H15NO4/c1-2-10-3-4-11-5-6-12-7(8)9/h2-6H2,1H3,(H2,8,9)
Inchi Key LWKFNWYUYUAOSC-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-153160

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-153160
Availability details for
Quantity
  • Availability
  • Add to Cart