ENA413170661 Enamine

4-(4-methyl-1,3-thiazol-5-yl)-1H-pyrrole-3-carboxylic acid

  • MDL number MFCD21294096
  • Cas Number : 1484948-42-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-153209

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-153209
Availability details for
Quantity
  • Availability
  • Add to Cart