ENA413170667 Enamine

4-amino-N,N,5-trimethyl-1H-pyrazole-3-carboxamide

  • MDL number MFCD16669740
  • Cas Number : 1340485-29-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-153258

Safety Information

msds https://www.enaminestore.com/catalog/EN300-153258
Availability details for
Quantity
  • Availability
  • Add to Cart