ENA413170668 Enamine

5-chloro-2-(cyclohexylmethoxy)benzene-1-sulfonyl chloride

  • MDL number MFCD17440085
  • Cas Number : 1545142-79-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-153261

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-153261
Availability details for
Quantity
  • Availability
  • Add to Cart