ENA413170675 Enamine

1-(oxolan-2-yl)prop-2-yn-1-amine

  • MDL number MFCD21264145
  • Cas Number : 1495393-47-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-153284

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-153284
Availability details for
Quantity
  • Availability
  • Add to Cart