ENA413170680 Enamine

ethyl 2-hydroxy-5-phenylpentanoate

  • MDL number MFCD21331371
  • Cas Number : 214336-45-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-153292

Safety Information

msds https://www.enaminestore.com/catalog/EN300-153292
Availability details for
Quantity
  • Availability
  • Add to Cart