ENA413170700 Enamine

1-(pyridazin-3-yl)piperidin-4-ol

  • MDL number MFCD16767815
  • Cas Number : 1217272-21-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-153318

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-153318
Availability details for
Quantity
  • Availability
  • Add to Cart